Learn More
Thermo Scientific™ Savant™ SpeedVac™ Medium Capacity Vacuum Concentrators for Combinatorial Chemistry Applications
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
Brand: Thermo Scientific™ SPD140P1-230
29055.06 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 161.00000kg
Description
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
SpeedVac SPD140DDA is a medium capacity vacuum concentrator used with a wide variety of solvents and offer resistance to TFA, DSMO and other aggressive solvents used in combinatorial chemistry applications. It is available in kit for ease of ordering.
SPD140DDA Features:
- All the product features of SPD130DLX PLUS:
- Radiant lamps within the chamber
- Heated lid for processing high boiling points (DMSO,DMF)
- Independent timers for both heat and run
- Audible alarm to alert user when operation is finished
- Built-in vacuum gauge and vacuum display
- Wider vapor port and tubing for enhanced vapor throughput - reducing drying times
- Adjustable vacuum level for preventing bumping of difficult solvent mixtures and optimizing drying times
- -5°C Cold Trap specially designed for trapping high boiling point solvents
Solvent and Solvent Combinations for SPD140P1 KIT
- Acetic acid
- Ammonium hydroxide
- Formic acid
- Hydrochloric acid
- TFA
- Methylene chloride and TFA
- Acetone and HCL
- Methylene chloride and MEOH
- DMSO and methanol
Solvent Combinations for SPD140 P2 KIT
- DMSO
- DMF
- Toluene
SPD140 P1 includes:
- Acid-Resistant SpeedVac™ Vacuum Concentrator
- -104°C Ultra-Low Temperature Refrigerated Vapor Trap
- Oil-Free ETFE/PFA 4-Headed Pump
- Rotor RH40-12, 12x 75mm holds 40
- Glass Condensation Flask
- Tubing and Fitting Kit
- CryoCool™ Heat Transfer Fluid (1L)
SPD140P2 includes:
- Acid-Resistant SpeedVac™ Vacuum Concentrator
- -50°C Universal Vacuum Source
- 24 x 1 dram vials (15 x 45 mm) 4ml - dram vial Rotor (RH24-15)
- Chemical Trap
- Tubing and Fitting Kit
- Disposable Cartridge
- Glas Condensation Flask
- Vacuum Pump Lubricating Oil
- CryoCool™ Heat Transfer Fluid (1L)
SPD140 P1 and P2 kits are recommended for:
- Solid phase extractions
- Drying lipid extractions
- Flash chromatography fractions
- Solid phase synthesis cleavage solutions such as protein hydrolyzates and evaporating synthesis solids
- Compound management
- Evaporating dissolution solventsRotors are available for a wide variety of applications and sample formats. Please check the rotor list.
Typical applications SPD140DDA:
Requires:
- Drying lipid extraction
- Flash chromatography fractions
- Combinatorial Chemistry
- Solid phase synthesis cleavage solutions such as protein hydrolyzates and evaporating synthesis solids
- Removing ethyl acetate from samples for CAT assay
- Drug metabolites in solid phase extracts
- Analyte elution solvents for evaporation
Specifications
UL Listed to US and Canadian requirements. CE marked | |
SPD140 P1 Kit | |
SpeedVac SPD140 Vacuum Concentrator Kit | |
230 V 50 Hz |