Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Large Capacity Concentrators
SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.
Brand: Thermo Scientific™ RBA2-FLSK-100
Code : NEW
Additional Details : Weight : 0.01000kg
Description
SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
- Easy to use
- Multi-size tube holes accommodate different sample sizes
- Require no additional accessories
- Fit directly into your concentrator
Specifications
Flasks: 2 x 100mL round bottom, Large capacity models | |
SPD210, SPD2030, SPD300DDA (requires RAD200) | |
RBA2-FLSK-100 |
Rotor | |
Flasks | |
Traditional |