missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB 202190, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Description
Inhibits p38 kinase activity through competition with ATP
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 100 mg |
Description | Yellow |
Content And Storage | -30°C to -10°C |
For Use With (Application) | Inhibits p38 kinase activity in vivo through competition with ATP |
Formula Weight | 331.35g/mol |
Melting Point | 240°C to 243°C |
Solubility | Soluble in DMSO at 30mg/ml |
Product Type | Signal Transduction Reagent |
Form | Crystalline Solid |
InChI Key | NJNKPVPFGLGHPA-UHFFFAOYSA-N |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'