missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Professional Scott™ Essential Couch Cover

Economic and portable, lightweight roll, ideal for protecting couches in the healthcare industry.

Brand:  Kimberly-Clark™ Professional 7161

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 12555518

  • 180.00 € / 6 rolls
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

  • Strong, tear-resistant and durable
  • Compatible with dispenser 7056
Specifications

Specifications

Paper wipers
17.316 in.
44.4 X 52cm
Wrapped Roll
44.4 cm
White
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers