missing translation for 'onlineSavingsMsg'
Learn More

Selenium(IV) oxide, 99.8%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number 11964121
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
7446-08-4
O2Se
110.97
MFCD00003562
JPJALAQPGMAKDF-UHFFFAOYSA-N
selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
24007
O=[Se]=O
This item is not returnable. View return policy
7446-08-4
O2Se
110.97
MFCD00003562
JPJALAQPGMAKDF-UHFFFAOYSA-N
selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
24007
O=[Se]=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 315.0°C
Color White to Beige
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 70 to 71.5% Se
Linear Formula SeO2
Fieser 01,992; 02,360; 03,245; 04,422; 05,575; 06,509; 08,439; 09,409; 10,345; 11,96; 13,272; 14,177; 15,89; 17,312
Merck Index 15,8571
Solubility Information Solubility in water: 38.4 g/100mL (14°C). Other solubilities: 1.11 parts/100 parts acetic acid (13.9°C), 10.16 parts/100 parts methanol (11.8°C), 6.67 parts/100 parts 93% ethanol (14°C), 4.35 parts/100 parts acetone (15.3°C)
IUPAC Name selenium dioxide
Formula Weight 110.96
Percent Purity 99.8%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Selenium(IV) oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Selenium(IV) oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'