missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Green Nitrile Disposable Gloves

Catalog Number 15930057
26.41 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.


Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Developed with sustainability in mind due to the unique production process, the glove offers skin-friendly hand protection, reduces the risk of allergy and feels soft to the skin. Comfortable and strong tensile strength ensures stability and a pleasant wearing experience even over longer periods.

  • PPE Cat. III
  • AQL 1.5
  • Powder-free
  • Dispoable, non-sterile
  • Comfortable to wear, even over longer periods of time
  • Skin-friendly formulation: no chlorine, no natural rubber latex proteins, no rubber accelerators
  • Manufactured while saving on water and energy, and reducing CO2 emissions
  • Synthetic inner coating
  • Textured fingertips provide extra grip
  • Stability and pleasant wearing experience
  • Glove material is resistant to blood and other body fluids, and protects against bacteria, fungi and viruses including SARS-CoV-2, Hepatitis B, Hepatitis C, and HIV
  • Ideal for applications including Personal Protection, Food/Catering, Consumer/Retail, Beauty/Body Art and Hairdressing
Certifications/Compliance (Europe) PPE Category III, EN 420, EN ISO 374-1, EN 374-2, EN 16523-1, EN 374-4, EN ISO 374-5, EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 2859-1, ASTM D6319, ASTM F1671
Chlorinated No
Accelerator Free Yes
Applications Industrial
AQL 1.5
Color Mint Green
Disposable Single-use
Exterior Finish Fingertip Textured
Material Nitrile
Powdered Powder Free
Size X-Small
Sterility Non-sterile
Glove Cuff Style Rolled
Series Semperguard™ Green
Tensile Strength 6 Newton
Length (Metric) 24 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'