missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Sempercare Shine+ Nitrile Examination Gloves

Catalog Number 15899047
17.24 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.


Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

A longer cuff offers users additional safety by covering wrists and underarms. Includes innovative foil opening for considerably easier glove removal.

  • The optimized length of the fingers significantly improves the wearing comfort
  • Powder-free natural latex examination glove excels by its extraordinary, well-gliding surface.
  • The textured fingers provide a good grip which is indispensable for safe instrument handling.
  • The synthetic inner coating achieved by the special Sempermed technology makes it easy and fast to don.
Certifications/Compliance ASTM, EN 455, EN 420, EN 374-1, EN 374-3
Classification CE Class I, CE Category III
Color White
Disposable Single-use
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Medium
Sterility Non-sterile
Thickness (Metric) 0.15 mm
For Use With (Application) Hospital
Glove Cuff Style Extended
Inspected for Medical Use Yes
Packaging Type Dispenser Box
Quantity 200/Pk.
Resistant To Chemical
Length (Metric) 26.5 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'