missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Vinyl Powdered Disposable Gloves

Catalog Number 15573516
10.63 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.


Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Semperguard™ Vinyl Powdered Disposable Gloves are safe for use with fat-free foodstuffs.

  • Pre-powdered
  • Vinyl construction
  • Free from latex proteins and accelerants
  • Free from DOP
  • Suitable for fat-free food contact
  • CE Category I minimal risks
  • Transparent in color
Certifications/Compliance ISO 9001, ISO 13485
Classification CE Category I
Cut Style Rolled
Color Transparent
Disposable Single-use
Latex Free Yes
Material Vinyl
Powdered Powdered
Product Type Disposable Gloves
Size Small
Thickness (Metric) 0.15 mm
For Use With (Application) Personal Protection, Professional Cleaning/Hygiene, Alternate Care
Glove Cuff Style Rolled
Packaging Type Dispenser Box
Quantity 100 Pk.
Length (Metric) 240 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'