Learn More
Robu™ Series 15 Filter Discs
Robu™ Series 15 Filter Discs feature a unique porous structure, which make them flexible and incredibly versatile for a variety of applications.
Brand:  Robu™ 15201
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12010412
  • 40.95 € / Pack of 10
Estimated Shipment: 10-07-2024
to see stock.
Add to basket
Description
Description
  • Surfaces untreated
  • Ground cylindrical
  • The faces are press molded and may show thickness deviations
Specifications
Specifications
Untreated surface
100 to 160 μm
3 ± 0.7 mm
20 mm
Glass
Cylindrical
Filter Disc