missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Series 16 0 Porosity Biplane Filter Disc

Brand:  Robu™ 16110

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10164261

  • 80.80 € / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

1Pack
Filter Disc
6.8 mm
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers