missing translation for 'onlineSavingsMsg'
Learn More

SGE™ ConnecTite™ Liners for Agilent™ GC

Designed for Agilent 7890, 6890, 6850, HP4890, 5890

Brand:  SGE™ 092326

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15269013

  • 343.00 € / Pack of 5


Estimated Shipment
19-08-2020
Add to basket

Description

Description

  • Range covers all applications and all popular instrument makes/ models
  • Range covers vaporizing and cool on-column injection techniques
  • Inlet liners ensure complete transfer of sample from syringe to the column without loss
  • 6.3mm OD, 4mm ID; 78.5mm length

Specifications

Specifications

Bottom hole
5/Pk.
0.4 cm
Agilent 7890, 6890, 6850, HP4890, 5890
Connectite Standard Liner
7.85 cm
0.63 cm
Videos
Documents