Learn More
Trajan™ Diamond MS Shimadzu Autosampler Syringes
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
Brand: Trajan™ 002780
Code : NEW
Additional Details : Weight : 0.02340kg
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Shimadzu instruments
Specifications
Diamond MS Autosampler Syringe | |
Ea. | |
Removable |
10R-S-GT-MS1 | |
10 μL |