Learn More
Trajan™ Thermo Scientific AS3000, AS2000 and AS200/800GC Autosampler Syringe
Fixed needle autosampler syringes. For reduced non specific sample interaction and increased analyte recovery
60.00 € - 351.00 €
Specifications
Needle Gauge | 26 G |
---|---|
Diameter (Metric) Outer | 0.47 mm |
Product Type | AS3000, AS2000 and AS200/800GC Autosampler Syringe |
Volume (Metric) | 10 μL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16682572
|
Trajan™
0029801 |
1/Pk. |
60.00 €
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15268463
|
Trajan™
0029861 |
6/Pk. |
351.00 €
Each |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13425072
|
Trajan™
0029801 |
1/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Thermo Scientific instruments
Specifications
26 G | |
0.47 mm | |
AS3000, AS2000 and AS200/800GC Autosampler Syringe | |
10 μL |