Learn More
Epredia™ Shandon™ Colormark™ Plus Slides
Improve cellular adhesion with positively charged Epredia™ Shandon™ ColorMark™ Plus Slides.
Brand: Epredia™ B9992010CS
Code : NEW
Additional Details : Weight : 1.00000kg
Description
Available in a variety of colors.
Specifications
Microscope Slide | |
Glass | |
Pink | |
144/Pk. |