Learn More
Epredia™ Single Cytofunnel™ with White Filter Cards
Hold sample volumes of up to 0.5 mL with the Epredia™ Single Cytofunnel™ with White Filter Cards.
Brand:  Epredia™ 1102548
Code : 50
Additional Details : Weight : 4.14000kg
 View more versions of this product
Product Code. 13127963
  • 865.00 € / Pack of 500
Estimated Shipment: 02-05-2024
to see stock.
Add to basket
Description
Description
  • Disposable sample chambers with white filter cards and caps
  • Pre-attached filter card
Specifications
Specifications
White
500/Cs.
0.1 to 0.5 mL
Single-use
Single Cytofunnel™, With Filter Cards and Caps