missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sharpsafe™ Sharps Bin, Cytotoxic and Cytostatic Waste for Incineration
The ideal combination of cost effectiveness and portability in medium usage areas.
12.20 €
Specifications
Certifications/Compliance | UN 3291, BS 7320 and French AFNOR NX30-500 Standards |
---|---|
Closure Color | Purple |
For Use With (Application) | Cytotoxic and cytostatic waste collection and incineration. |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15645697
|
Sharpsafe™
41202421 |
24 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Wide aperture facilitates easy disposal.
- Translucent lid, clear internal flap and visible fill line help ensure that the bin is not overfilled.
- In-mold label that cannot be removed.
- Simple four-click assembly.
- Large headroom.
- Inkjet-printed fill line and waste segregation message.
- Temporary and final closure feature
- Complies with UN 3291, BS 7320 and French AFNOR NX30-500 Standards
Specifications
UN 3291, BS 7320 and French AFNOR NX30-500 Standards | |
Purple | |
Cytotoxic and cytostatic waste collection and incineration. |