missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hospidex™ Sharpsafe™ Polypropylene Biohazard Container
Use the Sharpsafe™ Polypropylene Biohazard Container to transport dangerous biohazardous goods for disposal.
391.71 €
Specifications
Material | Polypropylene |
---|---|
Product Type | Biohazard Container |
Certifications/Compliance | NFX30-500 |
Autoclavable | Autoclavable |
For Use With (Application) | For collection of sharps waste treatment activities, pungent, sharp and / or pointed |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11988877
|
Hospidex™
SHA4160 |
1 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Yellow
- Polypropylene
- NFX 30-505 certification
Specifications
Polypropylene | |
NFX30-500 | |
For collection of sharps waste treatment activities, pungent, sharp and / or pointed |
Biohazard Container | |
Autoclavable |