missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sharpsafe™ Polypropylene Biohazard Sharps Container
Use the Sharpsafe™ Polypropylene Biohazard Sharps Container facilitate the safe management of wastes from healthcare activities.
Brand: Sharpsafe™ SHA4100
Code : A0
Additional Details : Weight : 14.62500kg
Description
- Polypropylene
- 50Pack
- Yellow
Specifications
4 L | |
248 mm | |
50 Pk. |
Yellow | |
175 mm | |
175 mm |