missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Shodex™ Asahipak ODP-50 C18 Mixed Mode HPLC Column, 5 μm, 4.6 mm x 250 mm
Carbon Load: 17%; Pore Size: 250 Å; Column Phase: Normal and Reversed Phase
Brand: Thermo Scientific™ F7620003
Code : NEW
Additional Details : Weight : 0.10000kg
Specifications
4.6 mm | |
Analysis of Amino Acids, Peptides and Proteins | |
250 mm | |
2 to 13 | |
L1 | |
Silica | |
Mixed Mode | |
Polymer Based | |
Reverse Phase HPLC |
5 μm | |
Octadecyl | |
5 μm | |
250 Å | |
C18 | |
17% | |
Normal and Reversed Phase | |
Column |