Learn More
Endecotts Sieve Shaker, Octagon 200 CL, 100-240 V, 50/60 Hz
Product Code. 15727649
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage unitSize
15727649 100/115/220/230 V Each
1 options
Product Code. 15727649 Supplier Endecotts Supplier No. OCT200CL/100240
Add to basket
Add to basket
Sieve Shaker Oct 200 CL
  • Octagon 200 CL sieve shaker with a measuring range from 20 μm to 125 mm, featuring electromagnetic 3D sieving motion, adjustable amplitude from 0 to 3 mm with digital setting in 0.1 mm increments, closed-loop amplitude control for consistent performance, and compatibility with 100 mm and 200 mm sieves as well as 3 in. and 8 in. sieve diameters.
Specifications
Load Bearing Capacity (Metric) 15 kg
Voltage 100/115/220/230 V