Learn More
Epredia™ Signature Series Cover Glass
Eliminate sticking and reduce waste with the uniquely manufactured Epredia™ Signature Series Cover Glass. Ideal for automatic coverslippers.
Brand:  Epredia™ 12460S
Code : LE
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12668066
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Thickness 1:
  • 0.13 - 0.17mm
Ordering Information:
  • Case of 10 Pk. (68-72/pack)
  • Available in several sizes
Specifications
Specifications
24 mm
#1 (0.13 to 0.16 mm)
Case of 10 Pk. (68-72/pack)
60 mm
60 x 24 mm