missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silica gel, preparative chromatography grade, spherical, 15 micron APS, 60 angstroms, 99.99+%

Brand:  Alfa Aesar™ 045824.14

Code : NEW

Additional Details : CAS Number : 63231-67-4 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15414985

  • 115.81 € / 25g


Estimated Shipment date
from Supplier 21-11-2018
Add to basket

Specifications

Chemical Identifiers

63231-67-4
78.098
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
H2O3Si
MFCD00011232
14768
O[Si](=O)O

Specifications

Silica gel
63231-67-4
≥99.99%
H2O3Si
15 Micron APS, 60Å
O[Si](=O)O
78.098
650m2/g
P.V. 1.00cc/g
Preparative Chromatography Grade
25g
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
14768
Spherical
Videos
SDS