Learn More
Macherey-Nagel™ Nano-SIL Unmodified HPTLC Silica Layers on Glass Plates
Separate mixtures with sharp separations, short developing times and short migration distances. Nano-SIL Unmodified HPTLC Silica Layers on Glass Plates have an increased detection sensitifvity compared to SIL G plates.
Brand: Macherey-Nagel™ 811012
Code : 16
Additional Details : Weight : 0.01000kg
Description
- Nano silica 60, specific surface (BET) ∼ 500 m2/g, mean pore size 60 Å, specific pore volume 0.75 mL/g, particle size 2–10 μm
- Indicator: manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
- Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents
Specifications
Silica 60 | |
60 Å | |
Glass Plate | |
0.75 mL/g | |
0.20 mm |
2 to 10 μm | |
25 Pk. | |
None | |
∼500 m2/g | |
10 x 10 cm |