missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide powder, coarse, 46 grit, Thermo Scientific Chemicals

Catalog Number p-7021933
Click to view available options
Quantity:
500 g
2500 g
10000 g
409-21-2
CSi
40.10
MFCD00049531
HBMJWWWQQXIZIP-UHFFFAOYSA-N
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
9863
CHEBI:29390
[C-]#[Si+]
409-21-2
CSi
40.10
MFCD00049531
HBMJWWWQQXIZIP-UHFFFAOYSA-N
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
9863
CHEBI:29390
[C-]#[Si+]

Silicon carbide is used in abrasives, in polishing and grinding. It is widely used in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. It compositely plays an important role in the crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon carbide is used in abrasives, in polishing and grinding. It is widely used in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. It compositely plays an important role in the crack-healing behavior.

Solubility
Soluble in molten sodium hydroxide, potassium hydroxide and in molten iron.

Notes
Keep it away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 500 g
Solubility Information Soluble in molten sodium hydroxide,potassium hydroxide and in molten iron.
IUPAC Name methanidylidynesilanylium
Formula Weight 40.1
Odor Odorless
Density 3.16
Chemical Name or Material Silicon carbide powder, coarse, 46 grit

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'