missing translation for 'onlineSavingsMsg'
Learn More

Silicon rod, 0.828cm (0.326in) dia, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 15466605
Click to view available options
Quantity:
100 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 7440-21-3
Molecular Formula Si
MDL Number MFCD00085311
Quantity 100 mm
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES [Si]
IUPAC Name silicon
Molecular Weight (g/mol) 28.09
PubChem CID 5461123
ChEBI CHEBI:27573
Percent Purity 99.999%
Odor Odorless
Form Rod
Assay (metals basis)
Chemical Name or Material Silicon rod, 0.828cm (0.326 in.) dia.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'