missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Silicone sealing ring, for GL threads, without PTFE washer

Seal for screw connection cap.

Brand:  DWK Life Sciences 286200405

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931541

  • 16.70 € / Pack of 10

Estimated Shipment: 21-12-2022
to see stock.
Add to basket

Description

Description

Suitable for PBT screw cap. Exemplary application: Fixing tubes

  • Suitable for PBT screw cap
Specifications

Specifications

Silicone
10/Pk.
Fixing tubes; Sealing
12 mm
GL 14
6 mm
Videos
SDS
Documents
Certificates
Special Offers

Special Offers