missing translation for 'onlineSavingsMsg'
Learn More

Deutsch Neumann™ Silicone Rubber Stoppers

For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.

Brand:  Deutsch Neumann™ 1010579

Additional Details : Weight : 0.41000kg

 View more versions of this product

Product Code. 10046401

  • 38.05 € / Each
Estimated Shipment: 29-04-2024
to see stock.
Add to basket

Description

Description

  • Acc. to DIN 12 871
  • Durometer 50 Shore A
  • Available in a wide range of sizes
Specifications

Specifications

Transparent
79 mm
Silicone
Stopper
92 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers