Learn More
Silver(I) fluoride, 99+%, ACROS Organics™
Brand: Acros Organics 210450050
Code : 29
Additional Details : CAS Number : 7775-41-9 Weight : 0.00500kg Transport : UN number : 3260 Chem class : 8 Pack group : II
Packaging | Plastic bottle |
---|---|
Quantity | 5g |
Description
C-X bond formation reagentChemical Identifiers
7775-41-9 | |
126.87 | |
REYHXKZHIMGNSE-UHFFFAOYSA-M | |
165912 | |
silver;fluoride |
AgF | |
MFCD00003410 | |
silver fluoride, silver 1+ fluoride, silver 1+ ion fluoride, argentous fluoride, ag-f, silverfluoride, silver salt, fluoride, silver fluoride,, silver i fluoride 5g | |
CHEBI:30340 | |
[F-].[Ag+] |
Specifications
Silver(I) fluoride | |
7775-41-9 | |
AgF | |
MFCD00003410 | |
15, 8653 | |
Solubility in water: in water: 182g/100mL (15.5°C) | |
[F-].[Ag+] | |
126.87 | |
CHEBI:30340 | |
Powder | |
5.8500g/mL | |
Solubility in water: in water: 182 g/100mL (15.5°C). | |
1150.0°C | |
435.0°C |
99+% | |
99% min | |
AgF | |
02,364; 05,581; 06,514; 15,287 | |
silver fluoride, silver 1+ fluoride, silver 1+ ion fluoride, argentous fluoride, ag-f, silverfluoride, silver salt, fluoride, silver fluoride,, silver i fluoride 5g | |
REYHXKZHIMGNSE-UHFFFAOYSA-M | |
silver;fluoride | |
165912 | |
126.87 | |
99+% | |
Plastic bottle | |
5.85 | |
Brown-Rust to Brown-Yellow | |
5g |
Safety and Handling
- Silver(I) fluoride
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.