missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver nanoparticles, 80nm, 0.02mg/mL, supplied in 2mM sodium citrate, 457nm absorption

Brand:  Alfa Aesar™ J67252.AC

Code : NEW

Additional Details : Weight : 0.00025kg

Quantity 25mL

 View more versions of this product

Product Code. 15474565

  • 77.30 € / 25mL

Estimated Shipment date
from Supplier 18-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
Silver nanoparticles applications include molecular diagnostics and photonic devices, which take advantage of the novel optical properties of these nanomaterials. An increasingly common application is the use of silver nanoparticles for antimicrobial coatings, and many textiles, keyboards, wound dressings, and biomedical devices now contain silver nanoparticles that continuously release a low level of silver ions to provide protection against bacteria.

Solubility
Fully miscible in water.

Notes
Light and air sensitive. Store away from heat, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]

Specifications

Silver nanoparticles
0.02mg/mL, supplied in 2mM sodium citrate, 457nm absorption
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
Odorless
23954
Liquid
Light Sensitive
25mL
7440-22-4
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
80nm
Yellow
Videos
SDS