Learn More
Simport™ Scientific M480 Disposable Embedding Cassettes
For holding and identifying tissue samples in processing, embedding and sectioning procedures during in vitro diagnostics
Brand:  Simport™ Scientific M480-12
Code : 04
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10428522
  • 155.00 € / Pack of 1500
Call for latest local availability
01 885 5854
Add to basket
Description
Description
  • Made of acetal polymer and resistant to chemical action of histological solvents
  • Fit securely in microtome chuck adapters
  • Accepts standard metal lids that keep specimens safe during processing
  • Slanted writing surface for easy sample identification
  • Available in three colors
Certifications
CE marked
Specifications
Specifications
Aqua
Acetal polymer
Embedding cassette
Videos