missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKF 83566 hydrobromide, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective D1-like dopamine receptor antagonist (Ki∽ 0.56 nM for D1; KB = 2 μM for D2). Also antagonist at the vascular 5-HT2 receptor (Ki = 11 nM). Displays selective inhibition of adenylyl cyclase 2 (AC2); inactive against AC1 or AC5. Centrally active following systemic administration in vivo.
Specifications
Quantity | 50 mg |
Formula Weight | 413.15 |
Percent Purity | >98% |
Chemical Name or Material | SKF 83566 hydrobromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'