missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC30A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563864
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: HSHGSRGAEY APLEEGPEEP LPLGNTSVR Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 83%.
Slc30a3 encodes a protein that is involved in accumulation of zinc in synaptic vesicles.Specifications
SLC30A3 | |
Polyclonal | |
Unconjugated | |
Slc30a3 | |
SLC30A3; solute carrier family 30 (zinc transporter), member 3; solute carrier family 30 member 3; solute carrier family 30 protein; zinc transporter 3; zinc transporter ZnT-3; ZNT3; znT-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7781 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99726 | |
Slc30a3 | |
Recombinant protein corresponding to Human SLC30A3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only