missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Slide Round Stirring Bar
The pivot ring reduces the contact surface and fosters optimal stir bar positioning
45.25 € - 47.50 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10366251
|
Kartell™
0078600 |
20, 57 mm |
45.25 €
Pack of 10 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10152231
|
Kartell™
0078700 |
24, 76 mm |
47.50 €
Pack of 10 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Description
Specifications
Magnet, PTFE | |
10 |