missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Safety-Walk™ Slip Resistant Conformable Tape, Series 500
Ideal for use on diamond grade safety plates, steps, and other irregular surfaces. Mineral coated and slip resistant, with aluminum foil backing. Conforms to irregular surfaces and around edges.
308.00 € - 925.00 €
Specifications
Adhesive Type | Pressure Sensitive Adhesive |
---|---|
For Use With (Application) | Light to Heavy Traffic Areas |
Product Code | Brand | Dimensions (L x W) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Color | Price | Quantity & Availability | ||||
17188670
|
3M™ Safety-Walk™
7100000611 |
18 x 0.051 m | Yellow |
308.00 €
Pack of 2 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
17118670
|
3M™ Safety-Walk™
7000126117 |
18.3 x 0.102 m | Black |
308.00 €
1 roll |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
17168600
|
3M™ Safety-Walk™
7000051948 |
18.3 x 0.305 m | Black |
925.00 €
1 roll |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
Description
- Anti-slip tape for irregular or contoured surfaces in light to heavy shoe-traffic areas
- Aluminum backing conforms to corners, curves, and irregular surfaces like diamond plating, riveted plates, and ladder rungs
- Suitable for application in dry, wet or oily environments
- Aggressive, high performance adhesive forms an excellent bond for a secure hold
- Tapes and treads resist wrinkling, curling, tearing, shrinking, and lifting
- Safety yellow color for high visibility and easy identification of hazard areas
- Custom shapes and sizes are also available
Specifications
Pressure Sensitive Adhesive | |
Light to Heavy Traffic Areas |