missing translation for 'onlineSavingsMsg'
Learn More

SCHOTT AG Lighting and Imaging™ Annular Fiber Optic Ringlight

Annular fiber optic ringlight. Mounts directly to 66 mm objectives by means of a thumb screw or objective adaptor. To use with Cold light source KL1500HAL, KL1600LED or KL2500LED

Brand:  SCHOTT AG Lighting and Imaging™ 157420

Additional Details : Weight : 5.00000kg

 View more versions of this product

Product Code. 10014580

  • 590.00 € / Each

Estimated Shipment: 22-05-2023
to see stock.
Add to basket

Description

Description

  • Ringlight with eco-friendly Puravis fiber optics
  • Fiber bundle protected with flexible metal PVC sheathing
  • Housing made of rugged, black anodized aluminum
  • Flexible length: 1000mm
Specifications

Specifications

Fiber Optic Ringlight
Cold Light Sources KL1600LED, KL2500LED or KL1500HAL
Eco-friendly Puravis Glass Optical Fibers
66 mm ID
1000 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers