missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences pDSG-IBA Small Mammalian Expression Vector
Designed for protein expression for mammalian HEK293E cells (MEXi 293E cells). Small Mammalian Expression Vector is a small transient expression vector especially developed for the use in combination with the MEXi mammalian expression system.
220.00 € - 266.00 €
Specifications
Constitutive or Inducible System | Constitutive |
---|---|
Marker Gene | Ampicillin |
Promoter | CMV |
Product Type | Vector |
Selection Agent (Eukaryotic) | Geneticin™ (G-418) |
Product Code | Brand | Protein Tag | Vector | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Protein Tag | Vector | Price | Quantity & Availability | ||||
15603888
|
IBA Lifesciences
5-5214-001 |
Strep-tag II, His Tag (6x) | pDSG-IBA45 |
266.00 €
5µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15613888
|
IBA Lifesciences
5-5219-001 |
Twin-Strep-tag | pDSG-IBA102 |
266.00 €
5µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15623888
|
IBA Lifesciences
5-5220-001 |
Twin-Strep-tag | pDSG-IBA103 |
266.00 €
5µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15633888
|
IBA Lifesciences
5-5221-001 |
Twin-Strep-tag | pDSG-IBA104 |
266.00 €
5µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15643888
|
IBA Lifesciences
5-5222-001 |
Twin-Strep-tag | pDSG-IBA105 |
266.00 €
5µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15663878
|
IBA Lifesciences
5-5200-001 |
Untagged | pDSG-IBAwt1 |
266.00 €
5µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15673878
|
IBA Lifesciences
5-5201-001 |
Untagged | pDSG-IBAwt2 |
266.00 €
5µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15693878
|
IBA Lifesciences
5-5211-001 |
His Tag (6x), Strep-tag II | pDSG-IBA43 |
266.00 €
5µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15683878
|
IBA Lifesciences
5-5209-001 |
His Tag (6x) | pDSG-IBA33 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Designed for protein expression for mammalian HEK293E cells (MEXi 293E cells). IBA Lifesciences GmbH Small Mammalian Expression Vector is a small transient expression vector especially developed for the use in combination with the MEXi mammalian expression system.Specifications
Constitutive | |
CMV | |
Geneticin™ (G-418) | |
5 μg |
Ampicillin | |
Vector | |
Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage |