Learn More
Thermo Scientific™ SMART Digest™ Trypsin Kits
Obtain fast, simple, and highly reproducible trypsin protein digestion for peptide characterization and quantitation in biopharmaceutical protein research.
Brand: Thermo Scientific™ 60109-101
Code : NEW
Additional Details : Weight : 0.04000kg
Description
Obtain high quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Kits. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.
Trypsin Protein DigestionSMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses trypsin, which cleaves peptides on the C-terminal side of lysine and arginine amino acid residues.
Easy and simple- Significantly increased speed of sample processing
- Simple process
- Easily automate entire protocol for high-throughput processing
- Increased reproducibility over existing protocols
- Higher sensitivity
- Higher data confidence
Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.
Cleaner SamplesReduction in the number of chemicals used and the immobilized trypsin design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. This results in results which are easier to interpret.
Couple with SOLAμ SPE Plates for DesaltingWhen used with the SMART Digest kits for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.
Specifications
Protein Digestion | |
SMART Digest Resin Pre-filled into PCR Tubes | |
C-terminal side of lysine and arginine amino acids | |
SMART Digest | |
SMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate |
SMART Digest Trypsin Kit, with Collection plate | |
Trypsin | |
None | |
Peptide Quantitation, Peptide Mapping |
Safety and Handling
- Trypsin Kits
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sodium azide, calcium chloride