missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Smooth Surface Filter Discs Qualitative Filter Papers, Grade: 3 w
Used for analytical and technical purposes.
8.50 € - 57.50 €
Specifications
Format | Disc |
---|---|
Material | Cotton Linters, Refined Pulp |
Filtration Speed | 15μm/sec. |
For Use With (Application) | For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications |
Grade | 3 w |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10032441
|
Sartorius™
FT-3-308-270 |
270 mm |
35.50 €
Pack of 100 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
10245991
|
Sartorius™
FT-3-308-185 |
185 mm |
22.90 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10554191
|
Sartorius™
FT-3-308-320 |
320 mm |
57.50 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10659151
|
Sartorius™
FT-3-308-070 |
70 mm |
10.10 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10082401
|
Sartorius™
FT-3-308-125 |
125 mm |
11.30 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10317841
|
Sartorius™
FT-3-308-110 |
110 mm |
11.30 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10787042
|
Sartorius™
FT-3-308-150 |
150 mm |
16.10 €
Pack of 100 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
10114381
|
Sartorius™
FT-3-308-055 |
55 mm |
8.50 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10082371
|
Sartorius™
FT-3-308-090 |
90 mm |
11.30 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10419701
|
Sartorius™
FT-3-308-240 |
240 mm |
35.50 €
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- For routine analyses like clarification, determination of substances
- Discs with a center hole for technical applications
- Made of refined pulp and cotton linters with >95% α-cellulose content
- Smooth surface
- Wet-strengthened
Specifications
Disc | |
15μm/sec. | |
3 w | |
0.14 mm |
Cotton Linters, Refined Pulp | |
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications | |
100 Pack | |
Qualitative Filter Paper |