missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Chosen reagents are combined in the accurate proportions according to the formula and thoroughly blended for a uniform distribution. A sample of the blended formulation is used to prepare liquid. The combinations are mixed to dissolve components and then autoclaved at 121°C for 15 minutes.
Specifications
Specifications
| Certifications/Compliance | Molecular Biology Certified™ |
| Content And Storage | Store at room temperature. |
| Description | SOC |
| Form | Powder |
| Formulation | Per liter: 20 g Tryptone-B; 5 g Yeast Extract-B; 0.5 g NaCl; 5 g MgSO4-7H2O; 20 mM glucose. Small quantities of biological buffers have been added for proper pH adjustment. |
| For Use With (Application) | Recovery step of Escherichia coli competent cell transformations |
| Quantity | 227 g |
| Regulatory Status | RUO |
| Specimen Type | E. coli |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?