missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Soda Lime Glass Class B Pipets

Brand:  Poulten Graf™ R376/A/5

Code : 01

Additional Details : Weight : 0.01000kg

Product Code. 12344418

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Color coded to BS 3996.Tufftip treated to reduce chipping and breakage.
Specifications

Specifications

Yes
B
Yes
5 Pack
1 mL
To Deliver
Soda Lime Glass
0.01 mL
0.01 mL
Straight Pipette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers