missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Soda Lime Glass Rimless Test Tube
Store test reagents and material. Fisherbrand™ soda lime glass rimless test tubes come in different sizes to meet your needs.
16.95 € - 36.80 €
Specifications
Certifications/Compliance | ISO 4142 |
---|---|
Material | Soda Lime Glass |
Quantity | 100 Pack |
Rimmed | No |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
15842265
|
Fisherbrand™
|
12 mm | 100 mm |
16.95 €
Pack of 100 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
15882265
|
Fisherbrand™
|
16 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Soda Lime Glass
- Rimless
- Disposable
Specifications
ISO 4142 | |
Soda Lime Glass | |
100 Pack | |
No |