missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Tube
Use the Soda Lime Glass Culture Tube to view cell cultures in the laboratory. This disposable soda lime glass rim top tube is plain (unlabeled) and features a sturdy, uniform bottom.
Brand: DWK Life Sciences A404008000711
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Rimmed top
- Unlableled
- Clear soda lime glass
- Uniform bottom
- Disposable
Specifications
Round bottom | |
200Pack | |
Test Tube |
40 mm | |
8 mm |