missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate Acetic acid solution, Volumetric, 0.1 M CH3COONa (in acetic acid), Honeywell Fluka™
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
CAS | 64-19-7 |
Quantity | 1 L |
Molecular Formula | C2H4O2 |
MDL Number | MFCD00036152 |
UN Number | UN2789 |
Beilstein | 3786729 |
Flash Point | 40°C (104°F) |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
SMILES | CC(O)=O |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'