missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Trihydrate, Crystal, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Brand:  JT Baker 4009-06

Additional Details : CAS Number : 6131-90-4 Weight : 5.83000kg

Packaging Poly Pail
Quantity 5kg

 View more versions of this product

Product Code. 15548204

  • 298.00 € / Each

Call for latest local availability
01 885 5854

Please to purchase this item. Need a web account? Register with us today!


Description

Description

    For Liquid Chromatography and Molecular Biology applications
Specifications

Chemical Identifiers

6131-90-4
136.079
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
CHEBI:32138
CC(=O)[O-].O.O.O.[Na+]
C2H9NaO5
AYRVGWHSXIMRAB-UHFFFAOYSA-M
23665404
sodium;acetate;trihydrate

Specifications

Sodium Acetate
6131-90-4
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
CC(=O)[O-].O.O.O.[Na+]
136.079
CHEBI:32138
Crystals
Poly Pail
5kg
Trihydrate
C2H9NaO5
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium;acetate;trihydrate
23665404
136.08
Ultrapure Bioreagent
1L = 1.45kg
Videos
Documents