missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate Trihydrate, Extra Pure, SLR, Crystal, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
C2H9NaO5, CAS Number-6131-90-4, unii-4550k0sc9b, thomaegelin, plasmafusin, sodium acetate tn, sodium acetate usan:jan, tutofusin, sodium acetate usp:jan, natrium acetate-3-wasser, acetic acid, sodium salt, trihydrate, sodium acetate trihydrate, 5kg, CHEBI:32138, White, 136.08g/mol
Specifications
Melting Point | 58°C |
Color | White |
pH | 8 to 9.5 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 136.08g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'