missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Benzoate, N.F. - F.C.C., J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Specifications
Quantity | 2.5 kg |
Formula Weight | 144.11 |
Grade | FCC/NF |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.44kg |
Chemical Name or Material | Sodium Benzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'