missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate decahydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog Number p-4510385
Click to view available options
Quantity:
100 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2-1-6132
CNa2O3·10H2O
286.14
XYQRXRFVKUPBQN-UHFFFAOYSA-L
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
151402
disodium;carbonate;decahydrate
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2-1-6132
CNa2O3·10H2O
286.14
XYQRXRFVKUPBQN-UHFFFAOYSA-L
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
151402
disodium;carbonate;decahydrate
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 34.0°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99+%
Linear Formula Na2CO3·10H2O
Fieser 7322
Merck Index 158731
Solubility Information Solubility in water: 210g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Formula Weight 286.14
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic Bottle
Specific Gravity 1.46
Density 1.46 g/mL
Chemical Name or Material Sodium carbonate decahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'