Learn More
Sodium chloride, meets analytical specification of Ph.Eur, Fisher Chemical™
ClNa, CAS Number-7647-14-5, salt, sodium chloride nacl, halite, common salt, table salt, sodium chloride, dendritis, saline, purex, rock salt, 5kg, 1461 deg.C, CHEBI:26710, White, 58.44g/mol, FAPWRFPIFSIZLT-UHFFFAOYSA-M, sodium;chloride, 801 deg.C, 3477, Plastic powder jar, 5.0 to 8.0, Solid, 5234
Brand: Fisher Chemical S/3160/65
146.05 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 22
Additional Details : CAS Number : 7647-14-5 Weight : 5.00000kg
Description
ClNa, CAS Number-7647-14-5, salt, sodium chloride nacl, halite, common salt, table salt, sodium chloride, dendritis, saline, purex, rock salt, 5kg, 1461 deg.C, CHEBI:26710, White, 58.44g/mol, FAPWRFPIFSIZLT-UHFFFAOYSA-M, sodium;chloride, 801 deg.C, 3477, Plastic powder jar, 5.0 to 8.0, Solid, 5234
Specifications
1461°C | |
White | |
5.0 to 8.0 | |
5 kg | |
MFCD00003477 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
58.44g/mol | |
1mmHg 865 |
801°C | |
7647-14-5 | |
Solid | |
ClNa | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
Plastic powder jar |