missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride solution, 1.0M, Honeywell™
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
CAS | 7647-14-5 |
Quantity | 1 L |
Molecular Formula | ClNa |
Linear Formula | NaCl |
MDL Number | MFCD00003477 |
UN Number | NONH for all modes of transport |
Beilstein | 3534976 |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
SMILES | [Na+].[Cl-] |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'