Learn More
Sodium Dihydrogen Orthophosphate Dihydrate, Extra Pure, SLR, Crystal, Fisher Chemical™
Brand: Fisher Chemical S/3720/65
168.66 EUR valid until 2022-06-30
Use promo code "16496" to get your promotional price.
Code : 21
Additional Details : CAS Number : 13472-35-0 Weight : 5.00000kg
Packaging | Plastic keg |
---|---|
Quantity | 5kg |
Chemical Identifiers
13472-35-0 | |
156.006 | |
VBJGJHBYWREJQD-UHFFFAOYSA-M | |
23673460 | |
O.O.OP(=O)(O)[O-].[Na+] |
H6NaO6P | |
149209 | |
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
sodium;dihydrogen phosphate;dihydrate |
Specifications
13472-35-0 | |
60°C | |
5kg | |
149209 | |
VBJGJHBYWREJQD-UHFFFAOYSA-M | |
sodium;dihydrogen phosphate;dihydrate | |
23673460 | |
Solid |
White | |
4.5 | |
H6NaO6P | |
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
O.O.OP(=O)(O)[O-].[Na+] | |
156.006 | |
156.01g/mol | |
Plastic keg |